Invitrogen
Manufacturer Code:PA547650
Catalog # PIPA547650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse gp130 Gln23-Glu617 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD130; CD130 CDw130 GP130 GP130-RAPS IL6R-beta IL6ST; gp130; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6R-beta; IL-6RB; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; Oncostatin-M receptor subunit alpha
Gene Aliases: 5133400A03Rik; AA389424; BB405851; CD130; D13Ertd699e; gp130; Il6st
UniProt ID: (Mouse) Q00560
Entrez Gene ID: (Mouse) 16195
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.