Invitrogen
Manufacturer Code:PA5109856
Catalog # PIPA5109856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 678-857 of human IL6ST (NP_001177910.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5133400A03Rik AA389424 Ac1055 BB405851 CD130 CDw130 D13Ertd699e EGK_16499 gp130 GP130-RAPS IL-6 IL-6 receptor subunit beta Il-6rb IL6R-beta IL6ST Interleukin-6 receptor subunit beta Interleukin-6 signal transducer Membrane glycoprotein 130 Oncostatin-M receptor subunit alpha; CD130; CD130 antigen; CDw130; gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6R-beta; IL-6RB; interleukin receptor beta chain; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; membrane glycoprotein gp130; Oncostatin-M receptor subunit alpha
Gene Aliases: 5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CDW130; D13Ertd699e; GP130; IL-6RB; IL6ST
UniProt ID: (Human) P40189, (Mouse) Q00560
Entrez Gene ID: (Human) 3572, (Rat) 25205, (Mouse) 16195
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.