Invitrogen
Manufacturer Code:MA536188
Catalog # PIMA536188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JE54-46 |
Immunogen | Synthetic peptide within Human GOT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AL022787 aspartate aminotransferase 2 aspartate aminotransferase mitochondrial aspartate transaminase 2 ASPATA FABP-1 FABPpm FABP-pm fatty acid-binding protein FLJ40994 Glutamate oxaloacetate transaminase 2 glutamate oxaloacetate transaminase 2 mitochondrial Glutamate oxaloacetate transaminase 2 mitochondrial (aspartate aminotransferase 2) glutamatic-oxaloacetic transaminase 2 mitochondrial glutamic-oxaloacetic transaminase 2 glutamic-oxaloacetic transaminase 2 mitochondrial glutamic-oxaloacetic transaminase 2 mitochondrial (aspartate aminotransferase 2) Got2 Got-2 KAT4 KATIV KYAT4 Kynurenine aminotransferase 4 Kynurenine aminotransferase IV Kynurenine--oxoglutarate transaminase 4 kynurenine--oxoglutarate transaminase IV Maat mAspAT mitAAT mitochondrial aspartate aminotransferase plasma membrane fatty acid binding protein plasma membrane-associated fatty acid-binding protein Transaminase A; aspartate aminotransferase 2; Aspartate aminotransferase, mitochondrial; aspartate transaminase 2; FABP-1; FABPpm; Fatty acid-binding protein; Glutamate oxaloacetate transaminase 2; glutamate oxaloacetate transaminase 2, mitochondrial; Glutamate oxaloacetate transaminase 2, mitochondrial (aspartate aminotransferase 2); glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); Kynurenine aminotransferase 4; Kynurenine aminotransferase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine--oxoglutarate transaminase IV; mAspAT; mitochondrial aspartate aminotransferase; plasma membrane fatty acid binding protein; Plasma membrane-associated fatty acid-binding protein; Transaminase A
Gene Aliases: AL022787; ASPATA; FABP-pm; Got-2; GOT2; KAT4; KATIV; KYAT4; Maat; mAspAT; mitAAT
UniProt ID: (Human) P00505, (Mouse) P05202, (Rat) P00507
Entrez Gene ID: (Human) 2806, (Mouse) 14719, (Rat) 25721
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.