Invitrogen
Manufacturer Code:PA577990
Catalog # PIPA577990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human GOT2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aspartate aminotransferase 2; Aspartate aminotransferase, mitochondrial; aspartate transaminase 2; FABP-1; FABPpm; Fatty acid-binding protein; FLJ40994 KAT4 KATIV mitAAT GOT2 "aspartate aminotransferase mitochondrial " fatty acid-binding protein FABPpm glutamate oxaloacetate transaminase 2 kynurenine aminotransferase IV transaminase A mAspAT plasma membrane-associated fatty acid-binding protein FABP-1 "glutamic-oxaloacetic transaminase 2 mitochondrial (aspartate aminotransferase 2) " Glutamate oxaloacetate transaminase 2; glutamate oxaloacetate transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); Kynurenine aminotransferase 4; Kynurenine aminotransferase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine--oxoglutarate transaminase IV; mAspAT; mitochondrial aspartate aminotransferase; plasma membrane fatty acid binding protein; Plasma membrane-associated fatty acid-binding protein; Transaminase A
Gene Aliases: AL022787; FABP-pm; Got-2; GOT2; KAT4; KATIV; KYAT4; mAspAT; mitAAT
UniProt ID: (Human) P00505, (Mouse) P05202
Entrez Gene ID: (Human) 2806, (Mouse) 14719
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.