Novus Biologicals
Manufacturer Code:NBP297808AF594
Catalog # NB097868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human GOSR1 fragment. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; cis-golgi SNARE; golgi integral membrane protein 2; golgi integral membrane protein 2 golgi SNAP receptor complex member 128 kDa cis-Golgi SNARE p28 Golgi SNARE 28 kDa GOLIM2 GOS28cis-golgi SNARE GOS-28GOS28/P28 GS2828 kDa Golgi SNARE protein P28; Golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOS-28
Gene Aliases: GOLIM2; GOS-28; GOS28; GOS28/P28; GOSR1; GS28; P28
UniProt ID: (Human) O95249
Entrez Gene ID: (Human) 9527
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.