Abnova
Manufacturer Code:H00009527K
Catalog # 89-203-312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GOSR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human GOSR1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; cis-golgi SNARE; golgi integral membrane protein 2; Golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOLIM2 GOS-28 GOS28 GOS28/P28 GS28 P28; GOS-28
Gene Aliases: GOLIM2; GOS-28; GOS28; GOS28/P28; GOSR1; GS28; P28
UniProt ID: (Human) O95249
Entrez Gene ID: (Human) 9527
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.