Novus Biologicals
Manufacturer Code:NBP276352
Catalog # NBP276352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Reacts with immunogen between amino acids 1 and 50 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Polyclonal Antibody
Protein Aliases: Coat protein GPP34; Coat protein GPP34 FLJ90675 Golgi phosphoprotein 3 golgi phosphoprotein 3 (coat-protein) golgi protein golgi-associated protein GOPP1 GPP34coat-protein MIDASgolgi peripheral membrane protein 1 34 kDa Mitochondrial DNA absence factor; coat-protein; golgi peripheral membrane protein 1, 34 kDa; Golgi phosphoprotein 3; golgi phosphoprotein 3 (coat-protein); golgi protein; golgi-associated protein; MIDAS; Mitochondrial DNA absence factor
Gene Aliases: GOLPH3; GOPP1; GPP34; MIDAS; Vps74
UniProt ID: (Human) Q9H4A6
Entrez Gene ID: (Human) 64083
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.