Invitrogen
Manufacturer Code:PA544628
Catalog # PIPA544628
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GOLGA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Golgi autoantigen, golgin subfamily a, 7; Golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; Golgin subfamily A member 7; golgin subfamily A member 7-like
Gene Aliases: AB041568; C130038N16Rik; GCP16; GOLGA3AP1; GOLGA7; GOLGA7A; HDCKB03P; HSPC041; MNCb-1213; R75586; wu:fc23b09; zgc:86753
UniProt ID: (Human) Q7Z5G4, (Bovine) Q5EA55, (Mouse) Q91W53, (Rat) Q6AYQ1
Entrez Gene ID: (Human) 51125, (Horse) 100050524, (Rabbit) 103352156, (Pig) 100156903, (Dog) 607476, (Bovine) 616809, (Mouse) 57437, (Guinea pig) 100724816, (Rat) 361171, (Zebrafish) 798622
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.