Abnova
Manufacturer Code:H00051125K
Catalog # 89-210-457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GOLGA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human GOLGA7 peptide using ARM Technology.
For Research Use Only
Protein Aliases: GCP16 GOLGA3AP1 GOLGA7A HSPC041 MGC21096 MGC4876; golgi autoantigen, golgin subfamily a, 7; Golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; Golgin subfamily A member 7
Gene Aliases: GCP16; GOLGA3AP1; GOLGA7; GOLGA7A; HDCKB03P; HSPC041
UniProt ID: (Human) Q7Z5G4
Entrez Gene ID: (Human) 51125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.