Invitrogen
Manufacturer Code:PA551495
Catalog # PIPA551495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GOLGA5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 31 protein; golgi autoantigen, golgin subfamily a, 5; golgi integral membrane protein 5; Golgin subfamily A member 5; Golgin-84; GOLIM5 RFG5 ret-II Golgin subfamily A member 5 RET-fused gene 5 protein cell proliferation-inducing gene 31 protein golgi autoantigen golgin subfamily a 5 golgi integral membrane protein 5 golgin-84; Protein Ret-II; Protein Sumiko; RET-fused gene 5 protein
Gene Aliases: GOLGA5; GOLIM5; PIG31; Ret-II; RETII; RFG5
UniProt ID: (Human) Q9UQQ7, (Mouse) Q9QYE6
Entrez Gene ID: (Human) 9950, (Mouse) 27277
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.