Abnova
Manufacturer Code:H00002796M11A
Catalog # 89-104-564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C2 |
Immunogen | GNRH1 (NP_00081624 a.a. &bsim 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GNRH1.
For Research Use Only
Protein Aliases: GNRH; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); Gonadotropin-releasing hormone I; GRH; leuteinizing-releasing hormone; LH-RH I; LHRH; LNRH; Luliberin I; Luteinizing hormone-releasing hormone I; Progonadoliberin I; Progonadoliberin-1; prolactin release-inhibiting factor
Gene Aliases: GNRH; GNRH1; GRH; LHRH; LNRH
UniProt ID: (Human) A0AVP0
Entrez Gene ID: (Human) 2796
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.