Novus Biologicals
Manufacturer Code:NBP297807FR
Catalog # NB047989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human GNPTG fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf27 c316G12.3 CAB56184 GlcNAc-1-phosphotransferase subunit gamma GNPTAGchromosome 16 open reading frame 27 LP2537 N-acetylglucosamine-1-phosphate transferase gamma subunit N-acetylglucosamine-1-phosphotransferase subunit gamma N-acetylglucosamine-1-phosphotransferase gamma subunit RJD9 UDP-N-acetylglucosamine-1-phosphotransferase subunit gamma; GlcNAc-1-phosphotransferase subunit gamma; N-acetylglucosamine-1-phosphate transferase, gamma subunit; N-acetylglucosamine-1-phosphotransferase subunit gamma; UDP-N-acetylglucosamine-1-phosphotransferase subunit gamma
Gene Aliases: C16orf27; CAB56184; GNPTAG; GNPTG; LP2537; RJD9
UniProt ID: (Human) Q9UJJ9
Entrez Gene ID: (Human) 84572
Molecular Function:
enzyme modulator
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.