Abnova
Manufacturer Code:H00084572B01P
Catalog # 89-017-187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | GNPTG (AAH14592 19 a.a. ~ 304 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human GNPTG protein.
For Research Use Only
Protein Aliases: C16orf27; GlcNAc-1-phosphotransferase subunit gamma; GNPTAG; LP2537; N-acetylglucosamine-1-phosphate transferase, gamma subunit; N-acetylglucosamine-1-phosphotransferase subunit gamma; RJD9; UDP-N-acetylglucosamine-1-phosphotransferase subunit gamma
Gene Aliases: C16orf27; CAB56184; GNPTAG; GNPTG; LP2537; RJD9
UniProt ID: (Human) Q9UJJ9
Entrez Gene ID: (Human) 84572
Molecular Function: enzyme modulator transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.