Invitrogen
Manufacturer Code:PA569636
Catalog # PIPA569636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human GNPTAB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GlcNAc phosphotransferase; GlcNAc-1-phosphotransferase subunits alpha/beta; glucosamine (UDP-N-acetyl)-lysosomal-enzyme N-acetylglucosamine phosphotransferase; GNPTA KIAA1208; N-acetylglucosamine-1-phosphate transferase alpha and beta subunits; N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits; N-acetylglucosamine-1-phosphotransferase subunit alpha; N-acetylglucosamine-1-phosphotransferase subunit beta; N-acetylglucosamine-1-phosphotransferase subunits alpha/beta; Stealth protein GNPTAB; UDP-N-acetylglucosamine-1-phosphotransferase subunits alpha/beta; UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosamine
Gene Aliases: EG432486; GNPTA; GNPTAB; ICD; KIAA1208; mKIAA1208; RGD1564821; si:ch211-234f20.3; wu:fd55d09; zgc:122985; zgc:158502
UniProt ID: (Human) Q3ZQK2, (Mouse) Q69ZN6, (Zebrafish) Q5RGJ8
Entrez Gene ID: (Horse) 100067011, (Rabbit) 100340567, (Bovine) 509610, (Dog) 475443, (Human) 79158, (Mouse) 432486, (Rat) 362865, (Guinea pig) 100716394, (Zebrafish) 553365
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.