Invitrogen
Manufacturer Code:PA531962
Catalog # PIPA531962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 184 of Human GNPNAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucosamine 6-phosphate N-acetyltransferase; GNPNAT Gpnat1 glucosamine 6-phosphate N-acetyltransferase phosphoglucosamine acetylase phosphoglucosamine transacetylase; Phosphoglucosamine acetylase; Phosphoglucosamine transacetylase
Gene Aliases: GNA1; GNPNAT; GNPNAT1; Gpnat1; RGD1563144
UniProt ID: (Human) Q96EK6
Entrez Gene ID: (Human) 64841, (Rat) 498486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.