Invitrogen
Manufacturer Code:PA5113303
Catalog # PIPA5113303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Glucosamine 6-phosphate N-acetyltransferase protein (1-184AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU017428 AU040593 EMeg32 Glucosamine 6-phosphate N-acetyltransferase glucosamine-6-phosphate acetyltransferase glucosamine-phosphate N-acetyltransferase 1 glucosamine-phosphate N-acetyltransferase 1 glucosamine-6-phosphate acetyltransferase glucosamine-phosphate N-acetyltransferase Gna1 GNPNAT Gnpnat1 Gpnat1 Gsnpat Phosphoglucosamine acetylase Phosphoglucosamine transacetylase Protein EMeg32 RGD1563144; Glucosamine 6-phosphate N-acetyltransferase; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase 1; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase; Phosphoglucosamine acetylase; Phosphoglucosamine transacetylase; Protein EMeg32
Gene Aliases: AU017428; AU040593; EMeg32; GNA1; GNPNAT; GNPNAT1; Gpnat1; Gsnpat
UniProt ID: (Human) Q96EK6, (Mouse) Q9JK38
Entrez Gene ID: (Human) 64841, (Mouse) 54342
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.