Novus Biologicals
Manufacturer Code:NBP30481420UL
Catalog # NB093421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-91 of human GNPNAT1 (NP_932332.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1.4 FLJ10607 glucosamine 6-phosphate N-acetyltransferase glucosamine-phosphate N-acetyltransferase 1 GNA1 GNPNAT Gpnat1 Phosphoglucosamine acetylase Phosphoglucosamine transacetylase; Glucosamine 6-phosphate N-acetyltransferase; Phosphoglucosamine acetylase; Phosphoglucosamine transacetylase
Gene Aliases: GNA1; GNPNAT; GNPNAT1; Gpnat1
UniProt ID: (Human) Q96EK6
Entrez Gene ID: (Human) 64841
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.