Novus Biologicals
Manufacturer Code:NBP299589R
Catalog # NB061889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GNPDA2. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.5.99.6 GlcN6P deaminase 2 glucosamine-6-phosphate deaminase 24921523I18Rik glucosamine-6-phosphate isomerase 2 Glucosamine-6-phosphate isomerase SB52 GNP2SB52 GNPDA 2 putative glucosamine-6-phosphate isomerase; glcN6P deaminase 2; Glucosamine-6-phosphate deaminase 2; Glucosamine-6-phosphate isomerase 2; Glucosamine-6-phosphate isomerase SB52; GNPDA 2
Gene Aliases: GNP2; GNPDA2; SB52
UniProt ID: (Human) Q8TDQ7
Entrez Gene ID: (Human) 132789
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.