Invitrogen
Manufacturer Code:MA537517
Catalog # PIMA537517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 237CT2.5.2 |
Immunogen | GNPDA1 recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: glcN6P deaminase 1; glcN6P deaminase 1 glucosamine-6-phosphate deaminase 1 Glucosamine-6-phosphate isomerase 1 glucose-6-phosphate isomerase GNP1 GNPDA GNPDA 1 GNPDA1 GNPI GPI HLN KIAA0060 oscillin; Glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; GNPDA 1; Oscillin
Gene Aliases: Gnp1; GNPDA; Gnpda1; Gnpi; oscillin
UniProt ID: (Mouse) O88958
Entrez Gene ID: (Mouse) 26384
Molecular Function:
hydrolase
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.