Invitrogen
Manufacturer Code:PA569532
Catalog # PIPA569532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human GNPDA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glcN6P deaminase 1; Glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; GNP1 GNPDA GNPI GPI HLN GNPDA 1 glcN6P deaminase 1 glucosamine-6-phosphate isomerase 1 oscillin; GNPDA 1; Oscillin
Gene Aliases: GNP1; GNPDA; GNPDA1; GNPI; GPI; HLN; KIAA0060; oscillin; zgc:110691
UniProt ID: (Human) P46926, (Bovine) A4FV08, (Mouse) O88958
Entrez Gene ID: (Horse) 100061298, (Rabbit) 100355853, (Dog) 478043, (Human) 10007, (Bovine) 522919, (Guinea pig) 100726426, (Mouse) 26384, (Rat) 683570, (Zebrafish) 550565
Molecular Function:
hydrolase
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.