Proteintech
Manufacturer Code:123121AP150UL
Catalog # 50-172-7883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GNPDA1 Fusion Protein Ag2961 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glcN6P deaminase 1; GlcN6P deaminase 1 GNPDA GNPDA 1 GNPDA1 GNPI GPI HLN KIAA0060 Oscillin; Glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; GNPDA 1; Oscillin
Gene Aliases: GNP1; GNPDA; GNPDA1; GNPI; GPI; HLN; KIAA0060; oscillin
UniProt ID: (Human) P46926, (Mouse) O88958
Entrez Gene ID: (Human) 10007, (Mouse) 26384, (Rat) 683570
Molecular Function:
hydrolase
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.