OriGene
Manufacturer Code:TA811117
Catalog # 50-168-3832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E7 |
Immunogen | Full length human recombit protein of human GNPAT produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:dihydroxyacetonephosphateacyltransferase; DAP-AT; DAP-AT DAPAT DHAPAT RCDP2; DHAP-AT; Dihydroxyacetone phosphate acyltransferase; Glycerone-phosphate O-acyltransferase
Gene Aliases: DAP-AT; DAPAT; DHAPAT; GNPAT; RCDP2
UniProt ID: (Human) O15228
Entrez Gene ID: (Human) 8443
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.