Invitrogen
Manufacturer Code:PA536447
Catalog # PIPA536447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 341-390 of human GNPAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acyl-CoA:dihydroxyacetonephosphate acyltransferase; Acyl-CoA:dihydroxyacetonephosphateacyltransferase; DAP-AT; DHAP-AT; Dihydroxyacetone phosphate acyltransferase; Dihydroxyacetone phosphate acyltransferase DAP-AT DHAP-AT Acyl-CoA:dihydroxyacetonephosphateacyltransferase Glycerone-phosphate O-acyltransferase GNPAT DAPAT DHAPAT; Glycerone-phosphate O-acyltransferase
Gene Aliases: AU019525; D1Ertd819e; DAP-AT; DAPAT; DHAPAT; GNPAT; RCDP2
UniProt ID: (Human) O15228, (Mouse) P98192, (Rat) Q9ES71
Entrez Gene ID: (Human) 8443, (Mouse) 14712, (Rat) 84470
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.