Abnova
Manufacturer Code:PAB22980
Catalog # 89-124-679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human GNL3L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant GNL3L.
For Research Use Only
Protein Aliases: FLJ10613; G protein nucleolar 3B; guanine nucleotide binding protein-like 3 (nucleolar)-like; Guanine nucleotide-binding protein-like 3-like protein; novel GTPase
Gene Aliases: GNL3B; GNL3L
UniProt ID: (Human) Q9NVN8
Entrez Gene ID: (Human) 54552
Molecular Function: G-protein enzyme modulator hydrolase nucleic acid binding signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.