Invitrogen
Manufacturer Code:PA539201
Catalog # PIPA539201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human GNG5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein gamma-5 subunit; GNGT5; guanine nucleotide binding protein (G protein), gamma 5; guanine nucleotide binding protein (G protein), gamma 5 subunit; guanine nucleotide binding protein gamma 5; Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-5
Gene Aliases: G(y)5; GNG5; GNGT5
UniProt ID: (Human) P63218, (Mouse) Q80SZ7, (Rat) P63219
Entrez Gene ID: (Human) 2787, (Mouse) 14707, (Rat) 79218
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.