Invitrogen
Manufacturer Code:PA5103875
Catalog # PIPA5103875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GNG2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 82 G gamma-I G protein subunit gamma 2 GBG2 Gng2 guanine nucleotide binding protein (G protein) gamma 2 guanine nucleotide binding protein (G protein) gamma 2 subunit guanine nucleotide binding protein gamma 2 guanine nucleotide-binding protein G(I)/G(O) gamma-2 subunit guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-2; G gamma-I; guanine nucleotide binding protein (G protein), gamma 2; guanine nucleotide binding protein gamma 2; guanine nucleotide-binding protein G(I)/G(O) gamma-2 subunit; Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-2
Gene Aliases: 82; GNG2
UniProt ID: (Human) P59768, (Mouse) P63213
Entrez Gene ID: (Human) 54331, (Mouse) 14702
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.