Invitrogen
Manufacturer Code:PA559797
Catalog # PIPA559797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GNB5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein, beta subunit 5L; G protein, beta-5 subunit; GB5 G protein beta subunit 5L G protein beta-5 subunit gbeta5 guanine nucleotide-binding protein subunit beta-5 guanine nucleotide-binding protein beta subunit 5L transducin beta chain 5; Gbeta5; guanine nucleotide binding protein (G protein), beta 5; Guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; Transducin beta chain 5
Gene Aliases: GB5; GNB5
UniProt ID: (Human) O14775
Entrez Gene ID: (Human) 10681
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.