Invitrogen
Manufacturer Code:PA5109781
Catalog # PIPA5109781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human GNB5 (NP_006569.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: flail; flailer; flr GB5 GBB5 Gbeta5 GBS GNB5 Guanine nucleotide-binding protein subunit beta-5 Transducin beta chain 5; G protein, beta subunit 5L; G protein, beta-5 subunit; Gbeta5; guanine nucleotide binding protein (G protein), beta 5; guanine nucleotide binding protein (G protein), beta polypeptide 5; guanine nucleotide binding protein, beta 5; Guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; guanine nucleotide-binding protein, beta-5 subunit; hug; Transducin beta chain 5
Gene Aliases: flr; GB5; Gbeta5; GBS; GNB5
UniProt ID: (Human) O14775, (Mouse) P62881, (Rat) P62882
Entrez Gene ID: (Human) 10681, (Mouse) 14697, (Rat) 83579
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.