Abnova
Manufacturer Code:H00010681A01
Catalog # 89-106-385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 90 a.a) partial recombinant protein with GST tag.,GNB5 (NP_057278 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant GNB5.
For Research Use Only
Protein Aliases: FLJ37457; FLJ43714; G protein, beta subunit 5L; G protein, beta-5 subunit; GB5; Gbeta5; guanine nucleotide binding protein (G protein), beta 5; Guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; Transducin beta chain 5
Gene Aliases: GB5; GNB5
UniProt ID: (Human) O14775
Entrez Gene ID: (Human) 10681
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.