Abnova
Manufacturer Code:PAB3270
Catalog # 89-130-919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GNB2L1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GNB2L1.
For Research Use Only
Protein Aliases: CDR62; Cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; Major Yo paraneoplastic antigen; Paraneoplastic cerebellar degeneration-associated antigen; Yo; Yo paraneoplastic antigen
Gene Aliases: CDR2; CDR62; PCD17; Yo
UniProt ID: (Human) Q01850
Entrez Gene ID: (Human) 1039
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.