Invitrogen
Manufacturer Code:PA572642
Catalog # PIPA572642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of DANRE gnb1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein beta-1 subunit beta subunit signal-transducing proteins GS/GI guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 1 guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1 transducin beta chain 1; guanine nucleotide binding protein (G protein), beta polypeptide 1; Guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1; Transducin beta chain 1
Gene Aliases: gnb1; wu:fb50b09; wu:fj94h04
UniProt ID: (Zebrafish) Q6PH57
Entrez Gene ID: (Zebrafish) 322104
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.