Invitrogen
Manufacturer Code:PA599154
Catalog # PIPA599154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Gz-alpha corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI847979 G protein subunit alpha z g(x) alpha chain GNAZ guanine nucleotide binding protein (G protein) alpha z polypeptide Guanine nucleotide binding protein alpha guanine nucleotide binding protein alpha z polypeptide guanine nucleotide binding protein alpha z subunit guanine nucleotide-binding protein G(z) subunit alpha GXA Gz Gz-alpha transducin alpha; G(x) alpha chain; guanine nucleotide binding protein (G protein), alpha z polypeptide; Guanine nucleotide binding protein alpha; guanine nucleotide binding protein, alpha z polypeptide; guanine nucleotide binding protein, alpha z subunit; Guanine nucleotide-binding protein G(z) subunit alpha; Gz-alpha; transducin alpha
Gene Aliases: AI847979; GNAZ; GXA; Gz
UniProt ID: (Human) P19086, (Mouse) O70443, (Rat) P19627
Entrez Gene ID: (Human) 2781, (Mouse) 14687, (Rat) 25740
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.