Novus Biologicals
Manufacturer Code:NBP120926
Catalog # NBP120926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KNQFLDLNLKKEDKE corresponding to internal region according to NP_001095856.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GDCA guanine nucleotide binding protein alpha transducing 3 guanine nucleotide-binding protein G(t) subunit alpha-3 gustducin Gustducin alpha-3 chain; guanine nucleotide binding protein, alpha transducing 3; Guanine nucleotide-binding protein G(t) subunit alpha-3; gustatory G protein; Gustducin alpha-3 chain; gustducin, alpha polypeptide
Gene Aliases: GDCA; GNAT3
UniProt ID: (Human) A8MTJ3
Entrez Gene ID: (Human) 346562
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.