Novus Biologicals
Manufacturer Code:NBP2840500.1ML
Catalog # NB031978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human GNAT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDCA guanine nucleotide binding protein alpha transducing 3 guanine nucleotide-binding protein G(t) subunit alpha-3 gustducin Gustducin alpha-3 chain; guanine nucleotide binding protein, alpha transducing 3; Guanine nucleotide-binding protein G(t) subunit alpha-3; gustatory G protein; Gustducin alpha-3 chain; gustducin, alpha polypeptide
Gene Aliases: GDCA; GNAT3
UniProt ID: (Human) A8MTJ3
Entrez Gene ID: (Human) 346562
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.