Invitrogen
Manufacturer Code:PA524553
Catalog # PIPA524553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 140-169 amino acids from the central region of human GNAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACHM4 GNATC cone-type transducin alpha subunit guanine nucleotide-binding protein G(t) subunit alpha-2 transducin alpha-2 chain transducin cone-specific alpha polypeptide; cone-type transducin alpha subunit; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; Guanine nucleotide-binding protein G(t) subunit alpha-2; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide
Gene Aliases: ACHM4; AW490837; Gnat-2; GNAT2; GNATC; Gt-2; Tcalpha
UniProt ID: (Human) P19087, (Mouse) P50149
Entrez Gene ID: (Human) 2780, (Mouse) 14686
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.