Invitrogen
Manufacturer Code:PA522340
Catalog # PIPA522340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 154 and 354 of GNAT2 (Uniprot IDNo.P19087) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cone-type transducin alpha subunit; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; Guanine nucleotide-binding protein G(t) subunit alpha-2; RP5-1160K1.4 ACHM4 GNATC cone-type transducin alpha subunit guanine nucleotide-binding protein G(t) subunit alpha-2 transducin alpha-2 chain transducin cone-specific alpha polypeptide; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide
Gene Aliases: ACHM4; AW490837; Gnat-2; GNAT2; GNATC; Gt-2; Tcalpha
UniProt ID: (Human) P19087, (Mouse) P50149
Entrez Gene ID: (Human) 2780, (Mouse) 14686
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.