Novus Biologicals
Manufacturer Code:NBP304537100UL
Catalog # NB095990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human GNAT2 (NP_005263.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACHM4guanine nucleotide-binding protein G(t) subunit alpha-2 cone-type transducin alpha subunit GNATC guanine nucleotide binding protein (G protein) alpha transducing activitypolypeptide 2 guanine nucleotide-binding protein G(t) alpha-2 subunit Transducin alpha-2 chain transducin cone-specific alpha polypeptide; cone-type transducin alpha subunit; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2; Guanine nucleotide-binding protein G(t) subunit alpha-2; Transducin alpha-2 chain; transducin, cone-specific, alpha polypeptide
Gene Aliases: ACHM4; GNAT2; GNATC
UniProt ID: (Human) P19087
Entrez Gene ID: (Human) 2780
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.