Proteintech
Manufacturer Code:551671AP150UL
Catalog # 50-173-6031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CSNBAD3 GBT1 GNAT1 GNATR Transducin alpha 1 chain; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1; Guanine nucleotide-binding protein G(t) subunit alpha-1; guanine nucleotide-binding protein G(T), alpha-1 subunit; rod-type transducin alpha subunit; Transducin alpha-1 chain; transducin, rod-specific
Gene Aliases: CSNB1G; CSNBAD3; GBT1; Gnat-1; GNAT1; GNATR; Ird1; Ird2; irdc; irdr; Tralpha; transducin
UniProt ID: (Human) P11488, (Mouse) P20612
Entrez Gene ID: (Human) 2779, (Mouse) 14685
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.