Invitrogen
Manufacturer Code:MA527115
Catalog # PIMA527115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A6 |
Immunogen | Full length human recombit protein of GNAS produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase stimulating G alpha proteinAHOAlternative gene product encoded by XL exonC20orf45dJ309F20.1.1dJ806M20.3.3Extra large alphas proteinGNASGNAS complex locusGNAS1GPSAGs alpha subunitGSAGSPGuanine nucleotide binding protein (G protein) alpha stimulating activity polypeptide 1Guanine nucleotide binding protein alpha stimulating activity polypeptide 1Guanine nucleotide binding protein G(s) subunit alpha isoforms shortGuanine nucleotide binding protein G(s) subunit alpha isoforms XLasGuanine nucleotide regulatory proteinMGC33735NESPNESP55Neuroendocrine secretory proteinPHP1APHP1BPHP1CPOHProtein ALEXSCG6Secretogranin VIXLalphasXLas; Adenylate cyclase-stimulating G alpha protein; Alternative gene product encoded by XL-exon; Extra large alphas protein; G protein subunit alpha S; GPIPIRRH peptide; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide regulatory protein; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; LHAL tetrapeptide; NESP55; neuroendocrine secretory protein; Neuroendocrine secretory protein 55; Protein ALEX; secretogranin VI; XLalphas
Gene Aliases: AHO; C20orf45; GNAS; GNAS1; GPSA; GSA; GSP; NESP; POH; SCG6; SgVI
UniProt ID: (Human) P84996
Entrez Gene ID: (Human) 2778
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.