Proteintech
Manufacturer Code:662531IG150UL
Catalog # 50-173-6686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A9C10 |
Immunogen | GNAS Fusion Protein Ag20530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase-stimulating G alpha protein; AHO C20orf45 GNAS GNAS complex locus GNAS1 GPSA GSA GSP NESP PHP1A PHP1B POH Protein ALEX; alpha-stimulatory subunit of GTP-binding protein; Alternative gene product encoded by XL-exon; Extra large alphas protein; G protein subunit alpha S; GPIPIRRH peptide; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide regulatory protein; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; guanine nucleotide-binding protein G(s), alpha subunit; guanine nucleotide-binding protein, alpha-stimulating activity polypeptide 1; LHAL tetrapeptide; NESP55; neuroendocrine secretory protein; Neuroendocrine secretory protein 55; Protein ALEX; secretogranin VI; XLalphas
Gene Aliases: AHO; C20orf45; GNAS; GNAS1; GPSA; GSA; GSP; NESP; NESP55; POH; PORGSA1; SCG6; SgVI
UniProt ID: (Human) P84996
Entrez Gene ID: (Human) 2778, (Pig) 100049657
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.