Invitrogen
Manufacturer Code:PA576679
Catalog # PIPA576679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GNAS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase stimulating G alpha proteinAHOAlternative gene product encoded by XL exonC20orf45dJ309F20.1.1dJ806M20.3.3Extra large alphas proteinGNASGNAS complex locusGNAS1GPSAGs alpha subunitGSAGSPGuanine nucleotide binding protein (G protein) alpha stimulating activity polypeptide 1Guanine nucleotide binding protein alpha stimulating activity polypeptide 1Guanine nucleotide binding protein G(s) subunit alpha isoforms shortGuanine nucleotide binding protein G(s) subunit alpha isoforms XLasGuanine nucleotide regulatory proteinMGC33735NESPNESP55Neuroendocrine secretory proteinPHP1APHP1BPHP1CPOHProtein ALEXSCG6Secretogranin VIXLalphasXLas; Adenylate cyclase-stimulating G alpha protein; Alternative gene product encoded by XL-exon; Extra large alphas protein; G protein subunit alpha S; G-alpha-8; GNAS; GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus; gnas {ECO:0000312|RGD:2716}; GPIPIRRH peptide; Gs alpha subunit; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide binding protein alpha s; guanine nucleotide regulatory protein; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; guanine nucleotide-binding protein G-s, alpha subunit; LHAL tetrapeptide; NESP55; neuroendocrine secretory protein; Neuroendocrine secretory protein 55; Protein ALEX; secretogranin VI; stimulatory G-protein alpha subunit; XLalphas
Gene Aliases: 5530400H20Rik; A930027G11Rik; AHO; C130027O20Rik; C20orf45; Galphas; GNAS; GNAS1; Gnasxl; Gnpas; GPSA; GSA; GSP; MNCb-5546; NESP; Nesp55; Nespl; Oed-Sml; Oedsml; P1; P2; P3; PHP1A; PHP1B; POH; SCG6; SgVI
UniProt ID: (Human) P84996, (Mouse) Q6R0H6, (Rat) P63095
Entrez Gene ID: (Human) 2778, (Mouse) 14683, (Rat) 24896
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.