Invitrogen
Manufacturer Code:PA542230
Catalog # PIPA542230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GNAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase-stimulating G alpha protein; alpha-stimulatory subunit of GTP-binding protein; Alternative gene product encoded by XL-exon; Extra large alphas protein; G protein alpha subunit; G protein subunit alpha S; G-alpha-8; GAIPIRRH peptide; GNAS; GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus; GNAS complex locus RP4-543J19.4 AHO C20orf45 GNAS1 GPSA GSA GSP MGC33735 PHP1A PHP1B POH dJ309F20.1.1 dJ806M20.3.3 NESP PHP1C; gnas {ECO:0000312|RGD:2716}; GPIPIRRH peptide; Gs alpha subunit; GTP-binding regulatory protein (G-s-alpha).; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide binding protein alpha s; guanine nucleotide regulatory protein; guanine nucleotide-binding protein G(s) subunit alpha isoforms; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; guanine nucleotide-binding protein G(s), alpha subunit; guanine nucleotide-binding protein G-s, alpha subunit; guanine nucleotide-binding protein Gs alpha1; guanine nucleotide-binding protein, alpha-stimulating activity polypeptide 1; LHAL tetrapeptide; LSAL tetrapeptide; NESP55; neuroendocrine secretory protein; Neuroendocrine secretory protein 55; Protein ALEX; secretogranin VI; stimulatory G-protein alpha subunit; unnamed protein product; XLalphas
Gene Aliases: 5530400H20Rik; A930027G11Rik; AHO; C130027O20Rik; C20orf45; G-SALPHA60A; Galphas; GNAS; GNAS1; Gnasxl; Gnpas; GPSA; GSA; GSP; MNCb-5546; NESP; NESP55; Nespl; Oed-Sml; Oedsml; P1; P2; P3; PHP1A; PHP1B; POH; PORGSA1; SCG6; SgVI
UniProt ID: (Human) P84996, (Bovine) O18979, (Mouse) Q6R0H6, (Rat) P63095
Entrez Gene ID: (Human) 2778, (Pig) 100049657, (Bovine) 281793, (Rabbit) 100008904, (Mouse) 14683, (Rat) 24896
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.