Proteintech
Manufacturer Code:101502AP150UL
Catalog # 50-172-6063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GNAS Fusion Protein Ag0197 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase-stimulating G alpha protein; AHO C20orf45 GNAS GNAS complex locus GNAS1 GPSA GSA GSP NESP PHP1A PHP1B POH; Alternative gene product encoded by XL-exon; Extra large alphas protein; G protein subunit alpha S; GPIPIRRH peptide; Gs alpha subunit; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1; guanine nucleotide regulatory protein; Guanine nucleotide-binding protein G(s) subunit alpha isoforms short; Guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas; LHAL tetrapeptide; NESP55; neuroendocrine secretory protein; Neuroendocrine secretory protein 55; Protein ALEX; secretogranin VI; stimulatory G-protein alpha subunit; XLalphas
Gene Aliases: 5530400H20Rik; A930027G11Rik; AHO; C130027O20Rik; C20orf45; Galphas; GNAS; GNAS1; Gnasxl; GPSA; GSA; GSP; MNCb-5546; NESP; Nespl; Oed-Sml; Oedsml; P1; P2; P3; PHP1A; PHP1B; POH; SCG6; SgVI
UniProt ID: (Human) P84996, (Mouse) Q6R0H6
Entrez Gene ID: (Human) 2778, (Mouse) 14683
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.