Invitrogen
Manufacturer Code:PA597796
Catalog # PIPA597796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse Guanine nucleotide-binding protein G(o) subunit alpha protein (2-354AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Go alpha; GTP-binding protein alpha o; GTP-binding protein alpha-o subunit; guanine nucleotide binding protein, alpha O polypeptide 1; Guanine nucleotide-binding protein G(o) subunit alpha
Gene Aliases: alphaO; AW050213; Galphao; Gna0; Gnao; Gnao1; RATBPGTPC
UniProt ID: (Rat) P59215, (Mouse) P18872
Entrez Gene ID: (Rat) 50664, (Mouse) 14681
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.