Invitrogen
Manufacturer Code:PA576657
Catalog # PIPA576657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GNAO1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686O0962G alpha oG protein alpha 0GalphaoGNAO 1GNAOGNAO&lowbarHUMANGNAO1Go alphaGuanine nucleotide binding protein (G protein) alpha activating activity polypeptide OGuanine nucleotide binding protein alpha activating polypeptideGuanine nucleotide binding protein G(o) subunit alphaGuanine nucleotide-binding protein G(o) subunit alpha; Go alpha; GO2-q chimeric G-protein; GTP-binding protein alpha-o subunit; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide O; Guanine nucleotide-binding protein G(o) subunit alpha; guanine nucleotide-binding regulatory protein 2
Gene Aliases: alphaO; AW050213; EIEE17; G-ALPHA-o; Galphao; Gna0; GNAO; GNAO1; HLA-DQB1
UniProt ID: (Human) P09471, (Mouse) P18872
Entrez Gene ID: (Human) 2775, (Mouse) 14681
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.