Invitrogen
Manufacturer Code:PA5101625
Catalog # PIPA5101625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GNAL corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610011C15Rik 9630020G10Rik Adenylate cyclase-stimulating G alpha protein olfactory type DYT25 G alpha 10 G protein subunit alpha L Galphaolf Gna10 GNAL Golf guanine nucleotide binding protein (G protein) alpha activating activity polypeptide olfactory type guanine nucleotide binding protein (G protein) alpha stimulating activity polypeptide olfactory type guanine nucleotide binding protein alpha 10 guanine nucleotide binding protein alpha stimulating olfactory type guanine nucleotide-binding protein G(olf) subunit alpha guanine nucleotide-binding protein G(olf) alpha subunit Olf olfa Olf-alpha protein (olfactory neuron-specific G protein) RGD1305940; Adenylate cyclase-stimulating G alpha protein, olfactory type; G alpha 10; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide, olfactory type; guanine nucleotide binding protein, alpha 10; Guanine nucleotide-binding protein G(olf) subunit alpha; guanine nucleotide-binding protein G(olf), alpha subunit; Olf-alpha protein (olfactory neuron-specific G protein)
Gene Aliases: 2610011C15Rik; 9630020G10Rik; DYT25; Galphaolf; Gna10; GNAL; Golf; Olf; RGD1305940
UniProt ID: (Human) P38405, (Rat) P38406, (Mouse) Q8CGK7
Entrez Gene ID: (Human) 2774, (Rat) 24611, (Mouse) 14680
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.