Invitrogen
Manufacturer Code:PA576885
Catalog # PIPA576885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GNAI3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G(i) alpha-3; guanine nucleotide binding protein (G protein), alpha inhibiting 3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; guanine nucleotide binding protein, alpha inhibiting 3; Guanine nucleotide binding, protein, alpha inhibiting polypeptide 3; Guanine nucleotide-binding protein G(i) subunit alpha-3; guanine nucleotide-binding protein G(k) subunit alpha; Guanine nucleotide-binding protein G(k) subunit alpha G(i) alpha-3 GNAI3
Gene Aliases: 87U6; AI158965; ARCND1; AW537698; Galphai3; GIP3A; Gnai-3; GNAI3
UniProt ID: (Human) P08754, (Rat) P08753, (Mouse) Q9DC51
Entrez Gene ID: (Human) 2773, (Rat) 25643, (Mouse) 14679
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.