Invitrogen
Manufacturer Code:PA527940
Catalog # PIPA527940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 68 and 354 of Human G protein alpha Inhibitor 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 87U6 g(i) alpha-3 guanine nucleotide-binding protein G(k) subunit alpha; G(i) alpha-3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; guanine nucleotide binding protein, alpha inhibiting 3; Guanine nucleotide-binding protein G(i) subunit alpha-3; guanine nucleotide-binding protein G(k) subunit alpha
Gene Aliases: 87U6; AI158965; ARCND1; AW537698; Galphai3; Gnai-3; GNAI3
UniProt ID: (Human) P08754, (Mouse) Q9DC51
Entrez Gene ID: (Human) 2773, (Mouse) 14679
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.