Novus Biologicals
Manufacturer Code:NBP2822370.1ML
Catalog # NB030166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human GNAI3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ26559 G(i) alpha-3 guanine nucleotide binding protein (G protein) alpha inhibiting activitypolypeptide 387U6 guanine nucleotide-binding protein G(k) subunit alpha; G(i) alpha-3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; Guanine nucleotide-binding protein G(i) subunit alpha-3
Gene Aliases: 87U6; ARCND1; GNAI3
UniProt ID: (Human) P08754
Entrez Gene ID: (Human) 2773
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.