Invitrogen
Manufacturer Code:PA5111148
Catalog # PIPA5111148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human GNAI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; Adenylate cyclase-inhibiting G alpha protein AU046200 BOS_4157 BPGTPB Gi Gialpha1 GNAI1 Gnai-1 Guanine nucleotide-binding protein G(i) subunit alpha-1; Gi1 protein alpha subunit; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; Guanine nucleotide-binding protein G(i) subunit alpha-1
Gene Aliases: Gi; GNAI1
UniProt ID: (Human) P63096
Entrez Gene ID: (Human) 2770
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.